Stampa…

4-Chloro-6-methoxypyrimidin-2-amine ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-4-chloro-6-methoxypyrimidine

A1520-25G A1520-5G
TCIAA1520-25GEA 76 EUR
TCIAA1520-25G TCIAA1520-5G
4-Chloro-6-methoxypyrimidin-2-amine ≥95.0% (by HPLC, titration analysis)
4-Chloro-6-methoxypyrimidin-2-amine
Formula: C₅H₆ClN₃O
Peso molecolare: 159,57 g/mol
MDL Number: MFCD00233535
CAS Number: 5734-64-5
EINECS: 410-050-9

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 168.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR