3-Aminophthalic acid ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A1516-1G
A1516-5G
TCIAA1516-1GEA
38
EUR
TCIAA1516-1G
TCIAA1516-5G
3-Aminophthalic acid ≥95.0% (by titrimetric analysis)
3-Aminophthalic acid
Formula:
C₈H₇NO₄ Peso molecolare: 181,15 g/mol |
CAS Number:
5434-20-8 EINECS: 226-595-9 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...