Stampa…

Acetoxime O-(2,4,6-Trimethylphenylsulphonate) ≥98.0% (by HPLC)

Fornitore: TCI

Synonyms: Acetone O-(2,4,6-Trimethylphenylsulphonyl)oxime, O-(2,4,6-Trimethylphenylsulphonyl)acetoxime

A1441-25G A1441-5G
TCIAA1441-25GEA 279 EUR
TCIAA1441-25G TCIAA1441-5G
Acetoxime O-(2,4,6-Trimethylphenylsulphonate) ≥98.0% (by HPLC)
Acetoxime O-(2,4,6-Trimethylphenylsulphonate)
Formula: C₁₂H₁₇NO₃S
MDL Number: MFCD01321127
CAS Number: 81549-07-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 94.0 to 98.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR