Allyl benzyl ether ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
((Allyloxy)methyl)benzene
A1427-100ML
A1427-25ML
TCIAA1427-100MLEA
244
EUR
TCIAA1427-100ML
TCIAA1427-25ML
Allyl benzyl ether ≥99.0% (by GC)
Allyl benzyl ether
Formula:
C₁₀H₁₂O Peso molecolare: 148,21 g/mol Punto di ebollizione: 205°C Flash Pt: 76°C |
MDL Number:
MFCD00078288 CAS Number: 14593-43-2 EINECS: 238-638-9 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...