Stampa…

9-Trifluoroacetylanthracene ≥98.0% (by GC)

Fornitore: TCI

A1422-1G A1422-5G
TCIAA1422-1GEA 57 EUR
TCIAA1422-1G TCIAA1422-5G
9-Trifluoroacetylanthracene ≥98.0% (by GC)
9-Trifluoroacetylanthracene
Formula: C₁₆H₉F₃O
MDL Number: MFCD00001258
CAS Number: 53531-31-0
EINECS: 258-609-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 86.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR