Methyl-5-aminolevulinate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl aminolevulinate hydrochloride, δ-Aminolevulinic Acid Methyl Ester Hydrochloride, 3-Aminoacetylpropionic Acid Methyl Ester Hydrochloride, 5-Amino-4-oxovaleric Acid Methyl Ester Hydrochloride, Methyl 5-amino-4-oxopentanoate hydrochloride, 5-Aminolevulinic Acid Methyl Ester Hydrochloride
Formula:
C₆H₁₁NO₃ Peso molecolare: 181,62 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00012868 CAS Number: 79416-27-6 EINECS: 279-151-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...