tert-Butyl acrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI
|
Pericolo
|
Synonyms:
Acrylic Acid tert-Butyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₇H₁₂O₂ Punto di ebollizione: 127°C Flash Pt: 19°C |
MDL Number:
MFCD00008809 CAS Number: 1663-39-4 EINECS: 216-768-7 UN: 2348 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...