N-Boc-1,6-diaminohexane ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-tert-Butoxycarbonyl-1,6-hexanediamine, N-Boc-1,6-Diaminohexane
A1375-1G
A1375-25G
A1375-5G
TCIAA1375-1GEA
25
EUR
TCIAA1375-1G
TCIAA1375-25G
TCIAA1375-5G
N-Boc-1,6-diaminohexane ≥98.0% (by titrimetric analysis)
N-Boc-1,6-diaminohexane
Formula:
C₁₁H₂₄N₂O₂ Peso molecolare: 216,33 g/mol Flash Pt: 125°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00671489 CAS Number: 51857-17-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...