Stampa…

Methylthiomethyl acetate ≥95.0% (by GC)

Fornitore: TCI
Attenzione

A1368-25G A1368-5G
TCIAA1368-25GEA 85 EUR
TCIAA1368-25G TCIAA1368-5G
Methylthiomethyl acetate ≥95.0% (by GC)
Methylthiomethyl acetate
Formula: C₄H₈O₂S
Punto di ebollizione: 62°C
Flash Pt: 44°C
MDL Number: MFCD00075528
CAS Number: 16437-69-7
UN: 3272
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR