Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate, 5-Amino-3-methyl-2,4-thiophenedicarboxylic Acid Diethyl Ester, 2-Amino-3,5-bis(ethoxycarbonyl)-4-methylthiophene
A1354-25G
A1354-5G
TCIAA1354-25GEA
55
EUR
TCIAA1354-25G
TCIAA1354-5G
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate ≥98.0% (by GC)
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate
Formula:
C₁₁H₁₅NO₄S Peso molecolare: 257,30 g/mol |
MDL Number:
MFCD00005450 CAS Number: 4815-30-9 EINECS: 225-388-0 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...