4-Amino-p-terphenyl ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[1,1:4,1-Terphenyl]-4-amine
A1340-1G
A1340-25G
A1340-5G
TCIAA1340-1GEA
61
EUR
TCIAA1340-1G
TCIAA1340-25G
TCIAA1340-5G
4-Amino-p-terphenyl ≥97.0% (by GC)
4-Amino-p-terphenyl
Formula:
C₁₈H₁₅N Peso molecolare: 245,33 g/mol |
MDL Number:
MFCD00191302 CAS Number: 7293-45-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...