Stampa…
4-Acetyl-4'-methylbiphenyl ≥98.0% (by GC)
Fornitore: TCI
A1322-5G A1322-1G
TCIAA1322-5GEA 163 EUR
TCIAA1322-5G TCIAA1322-1G
4-Acetyl-4'-methylbiphenyl ≥98.0% (by GC)
4-Acetyl-4'-methylbiphenyl
Formula: C₁₅H₁₄O
MDL Number: MFCD00191374
CAS Number: 5748-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be42177b562d29aea1311c0bdba9bcf33caa3c3&vid=0be42177b562d29aea1311c0bdba9bcf33caa3c3&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %