Stampa…

2-Acetyl-7-hydroxybenzofuran ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-(7-Hydroxybenzofuran-2-yl)ethan-1-one, 2-Acetyl-7-hydroxycoumarone

A1316-10G A1316-5G
TCIAA1316-10GEA 116 EUR
TCIAA1316-10G TCIAA1316-5G
2-Acetyl-7-hydroxybenzofuran ≥98.0% (by GC)
2-Acetyl-7-hydroxybenzofuran
Formula: C₁₀H₈O₃
Peso molecolare: 176,17 g/mol
MDL Number: MFCD00191373
CAS Number: 40020-87-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR