Stampa…

3-Acetyl-2-(methylaminosulphonyl)thiophene ≥98.0% (by GC)

Fornitore: TCI

A1315-5G A1315-1G
TCIAA1315-5GEA 381 EUR
TCIAA1315-5G TCIAA1315-1G
3-Acetyl-2-(methylaminosulphonyl)thiophene ≥98.0% (by GC)
3-Acetyl-2-(methylaminosulphonyl)thiophene
Formula: C₇H₉NO₃S₂
MDL Number: MFCD00191372
CAS Number: 138891-01-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR