Stampa…

5-Aminoisophthalic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A1290-25G A1290-500G A1290-100G
TCIAA1290-25GEA 16 EUR
TCIAA1290-25G TCIAA1290-500G TCIAA1290-100G
5-Aminoisophthalic acid ≥98.0% (by titrimetric analysis)
5-Aminoisophthalic acid
Formula: C₈H₇NO₄
Peso molecolare: 181,15 g/mol
MDL Number: MFCD00002522
CAS Number: 99-31-0
EINECS: 202-748-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0% (calcd.on anh.substance)
Water max. 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR