Stampa…

2-Amino-p-cresol hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-5-methylaniline Hydrochloride, 2-Amino-4-methylphenol Hydrochloride

A1276-25G A1276-500G
TCIAA1276-25GEA 24 EUR
TCIAA1276-25G TCIAA1276-500G
2-Amino-p-cresol hydrochloride ≥98.0% (by titrimetric analysis)
2-Amino-p-cresol hydrochloride
Formula: C₇H₉NO
Peso molecolare: 159,61 g/mol
MDL Number: MFCD00191600
CAS Number: 2977-71-1
EINECS: 221-026-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light red to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR