1-Acetoxy-2-methoxy-4-(1-propenyl)benzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Methoxy-4-(prop-1-en-1-yl)phenyl acetate, Isoeugenol acetate, Acetylisoeugenol
A1184-25G
A1184-500G
TCIAA1184-25GEA
16
EUR
TCIAA1184-25G
TCIAA1184-500G
1-Acetoxy-2-methoxy-4-(1-propenyl)benzene ≥98.0% (by GC)
1-Acetoxy-2-methoxy-4-(1-propenyl)benzene
Formula:
C₁₂H₁₄O₃ Peso molecolare: 206,24 g/mol Punto di ebollizione: 283°C |
MDL Number:
MFCD00026984 CAS Number: 93-29-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 83.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...