(3S)-(+)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine
A1174-10G
A1174-25G
TCIAA1174-10GEA
133
EUR
TCIAA1174-10G
TCIAA1174-25G
(3S)-(+)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Formula:
C₁₁H₁₆N₂ Peso molecolare: 176,26 g/mol |
MDL Number:
MFCD00082766 CAS Number: 114715-38-7 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +9.3 to +9.7 deg (C=5, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...