Stampa…

(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine

A1172-25G A1172-5G
TCIAA1172-25GEA 321 EUR
TCIAA1172-25G TCIAA1172-5G
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formula: C₉H₁₈N₂O₂
MDL Number: MFCD00143194
CAS Number: 122536-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66a3e039a2b51ec87d168b045dd5a88102d27609&vid=66a3e039a2b51ec87d168b045dd5a88102d27609&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Specific rotation [a]20/D -19.0 to -22.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR