Stampa…

(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate

A1171-25G A1171-5G
TCIAA1171-25GEA 246 EUR
TCIAA1171-25G TCIAA1171-5G
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formula: C₉H₁₈N₂O₂
Peso molecolare: 186,26 g/mol
MDL Number: MFCD00143191
CAS Number: 122536-77-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Specific rotation [a]20/D +19.0 to +22.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR