Stampa…

N-(2-Aminoethyl)glycine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-((2-Aminoethyl)amino)acetic acid, N-(2-Aminoethyl)-Gly-OH, Ethylenediamine-N-monoacetic Acid

A1153-1G A1153-100MG
TCIAA1153-1GEA 99 EUR
TCIAA1153-1G TCIAA1153-100MG
N-(2-Aminoethyl)glycine ≥97.0% (by titrimetric analysis)
N-(2-Aminoethyl)glycine
Formula: C₄H₁₀N₂O₂
Peso molecolare: 118,14 g/mol
MDL Number: MFCD00144824
CAS Number: 24123-14-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 141.0 to 145.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR