Angelic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(Z)-2-Methyl-2-butenoic Acid, 2-Methylisocrotonic Acid, (Z)-2-Methylbut-2-enoic acid
A1136-1G
A1136-5G
TCIAA1136-1GEA
76
EUR
TCIAA1136-1G
TCIAA1136-5G
Angelic acid ≥98.0% (by GC, titration analysis)
Angelic acid
Formula:
C₅H₈O₂ Peso molecolare: 100,12 g/mol Punto di ebollizione: 96°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002654 CAS Number: 565-63-9 EINECS: 209-284-2 UN: 3261 ADR: 8,III Merck Index: 14,00646 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...