Stampa…

3-(N-Acetyl-N-ethylamino)pyrrolidine ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

A1113-5G A1113-1G
TCIAA1113-5GEA 397 EUR
TCIAA1113-5G TCIAA1113-1G
3-(N-Acetyl-N-ethylamino)pyrrolidine ≥98.0% (by titrimetric analysis)
3-(N-Acetyl-N-ethylamino)pyrrolidine
Formula: C₈H₁₆N₂O
Peso molecolare: 156.23 g/mol
MDL Number: MFCD00059021
CAS Number: 115445-29-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR