4-Aminostyrene ≥98.0% (by titrimetric analysis) stabilizzato
Fornitore: TCI
|
Attenzione
|
Synonyms:
4-Vinylaniline
A1019-25G
A1019-5G
TCIAA1019-25GEA
890
EUR
TCIAA1019-25G
TCIAA1019-5G
4-Aminostyrene ≥98.0% (by titrimetric analysis) stabilizzato
4-Aminostyrene
Formula:
C₈H₉N Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00015329 CAS Number: 1520-21-4 EINECS: 216-185-8 |
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |