5-Amino-8-hydroxyquinoline dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Amino-8-quinolinol Dihydrochloride
A1005-25G
A1005-5G
TCIAA1005-25GEA
507
EUR
TCIAA1005-25G
TCIAA1005-5G
5-Amino-8-hydroxyquinoline dihydrochloride ≥98.0% (by HPLC, total nitrogen)
5-Amino-8-hydroxyquinoline dihydrochloride
Formula:
C₉H₈N₂O Peso molecolare: 233,09 g/mol |
MDL Number:
MFCD00012737 CAS Number: 21302-43-2 |
Risultati del test di specificazione
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...