Aspartame
Fornitore: TCI
A0997-1G
A0997-25G
TCIAA0997-1GEA
16
EUR
TCIAA0997-1G
TCIAA0997-25G
Aspartame
Proteine e Peptidi
- Specific rotation [a]20/D: +12,0 to +17,0° (C=4, 15 mol/L Formic acid)
- Water: Max. 4,5%
- EINECS: 245-261-3
- Merck Index_V14: 14,00839
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...