Stampa…

3-Methoxy-3-methylbutyl acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Acetic Acid 3-Methoxy-3-methylbutyl Ester

A0964-500ML A0964-25ML
TCIAA0964-500MLEA 66 EUR
TCIAA0964-500ML TCIAA0964-25ML
3-Methoxy-3-methylbutyl acetate ≥98.0% (by GC)
3-Methoxy-3-methylbutyl acetate
Formula: C₈H₁₆O₃
Peso molecolare: 160,21 g/mol
Punto di ebollizione: 74°C
Flash Pt: 73°C
MDL Number: MFCD00059340
CAS Number: 103429-90-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR