Stampa…
2-(Dimethylamino)ethyl acetate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: Acetic Acid 2-(Dimethylamino)ethyl Ester
A0929-25ML A0929-500ML
TCIAA0929-25MLEA 35 EUR
TCIAA0929-25ML TCIAA0929-500ML
2-(Dimethylamino)ethyl acetate ≥98.0% (by GC, titration analysis)
2-(Dimethylamino)ethyl acetate
Formula: C₆H₁₃NO₂
Peso molecolare: 131,18 g/mol
Punto di ebollizione: 152°C
MDL Number: MFCD00041912
CAS Number: 1421-89-2
EINECS: 215-827-4
UN: 1993
ADR: 3,III
Visualizza i codici
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %