Stampa…

6-Aminofluorescein (isomer II) ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Fluoresceinamine, 6-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, 6-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

A0864-1G A0864-5G
TCIAA0864-1GEA 150 EUR
TCIAA0864-1G TCIAA0864-5G
6-Aminofluorescein (isomer II) ≥95.0% (by titrimetric analysis)
6-Aminofluorescein (isomer II)
Formula: C₂₀H₁₃NO₅
Peso molecolare: 347,33 g/mol
MDL Number: MFCD00005051
CAS Number: 51649-83-3
EINECS: 257-334-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR