Stampa…

O-Acetyl-L-serine hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-3-Acetoxy-2-aminopropanoic acid hydrochloride, H-Ser(Ac)-OH.HCl

A0834-1G A0834-100MG
TCIAA0834-1GEA 119 EUR
TCIAA0834-1G TCIAA0834-100MG
O-Acetyl-L-serine hydrochloride ≥98.0% (by titrimetric analysis)
O-Acetyl-L-serine hydrochloride
Formula: C₅H₉NO₄
Peso molecolare: 183,59 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00060169
CAS Number: 66638-22-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +14.0 to +18.0 deg (C=5, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR