Stampa…

Acetylenedicarboxylic acid monopotassium salt ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Potassium Hydrogen Acetylenedicarboxylate, Monopotassium Acetylenedicarboxylate

A0830-25G A0830-500G
TCIAA0830-25GEA 24 EUR
TCIAA0830-25G TCIAA0830-500G
Acetylenedicarboxylic acid monopotassium salt ≥95.0% (by titrimetric analysis)
Acetylenedicarboxylic acid monopotassium salt
Formula: C₄H₂O₄
Peso molecolare: 152,15 g/mol
MDL Number: MFCD00013143
CAS Number: 928-04-1
EINECS: 213-169-2
UN: 2811
ADR: 6.1,I

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR