Stampa…

Amyl acetoacetate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Acetoacetic Acid Pentyl Ester, Acetoacetic Acid Amyl Ester, Pentyl Acetoacetate

A0811-25ML A0811-500ML
TCIAA0811-25MLEA 29 EUR
TCIAA0811-25ML TCIAA0811-500ML
Amyl acetoacetate ≥98.0% (by GC)
Amyl acetoacetate
Formula: C₉H₁₆O₃
Peso molecolare: 172,22 g/mol
Punto di ebollizione: 222°C
MDL Number: MFCD00059433
CAS Number: 6624-84-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR