Stampa…

Atropine ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (+/-)-Hyoscyamine, Tropine DL-tropate

A0754-25G A0754-5G
TCIAA0754-25GEA 435 EUR
TCIAA0754-25G TCIAA0754-5G
Atropine ≥99.0% (by titrimetric analysis)
Atropine
Formula: C₁₇H₂₃NO₃
Peso molecolare: 289,38 g/mol
MDL Number: MFCD00022622
CAS Number: 51-55-8
EINECS: 200-104-8
UN: 1544
ADR: 6.1,III
Merck Index: 14,00875

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 114.0 to 118.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR