Stampa…

3-Aminocyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: 3-Aminocyclohexane-1-carboxylic acid, 3-AminocyclohexanecarboxylicAcid, 3-Aminocyclohexanecarboxylic acid (cis- and trans- mixture)

A0749-25G A0749-5G
TCIAA0749-25GEA 222 EUR
TCIAA0749-25G TCIAA0749-5G
3-Aminocyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by titrimetric analysis)
3-Aminocyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₇H₁₃NO₂
MDL Number: MFCD00059562
CAS Number: 25912-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48517bc66c9e6a999a73099613b229721effddb9&vid=48517bc66c9e6a999a73099613b229721effddb9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR