Stampa…

Isobutyl acrylate ≥99.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: Acrylic Acid Isobutyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A0747-25ML A0747-500ML
TCIAA0747-25MLEA 18 EUR
TCIAA0747-25ML TCIAA0747-500ML
Isobutyl acrylate ≥99.0% (by GC) stabilizzato
Isobutyl acrylate
Formula: C₇H₁₂O₂
Peso molecolare: 128,17 g/mol
Punto di ebollizione: 132°C
Flash Pt: 31°C
MDL Number: MFCD00042865
CAS Number: 106-63-8
EINECS: 203-417-8
UN: 2527
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Acid Value max. 0.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR