Diisobutyl adipate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Adipic Acid Diisobutyl Ester
A0706-25ML
A0706-500ML
TCIAA0706-25MLEA
16
EUR
TCIAA0706-25ML
TCIAA0706-500ML
Diisobutyl adipate ≥99.0% (by GC)
Diisobutyl adipate
Formula:
C₁₄H₂₆O₄ Peso molecolare: 258,36 g/mol Punto di ebollizione: 293°C Flash Pt: 156°C |
MDL Number:
MFCD00053722 CAS Number: 141-04-8 EINECS: 205-450-3 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...