Stampa…

6-Amino-2-mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A0682-25G A0682-5G
TCIAA0682-25GEA 582 EUR
TCIAA0682-25G TCIAA0682-5G
6-Amino-2-mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)
6-Amino-2-mercaptobenzothiazole
Formula: C₇H₆N₂S₂
Peso molecolare: 182,26 g/mol
MDL Number: MFCD00059164
CAS Number: 7442-07-1
EINECS: 231-184-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR