Stampa…

7-Anilino-4-hydroxy-2-naphthalenesulphonic acid ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 7-Anilino-4-naphthol-2-sulphonic acid, Phenyl J acid

A0467-25G A0467-500G
TCIAA0467-25GEA 32 EUR
TCIAA0467-25G TCIAA0467-500G
7-Anilino-4-hydroxy-2-naphthalenesulphonic acid ≥95.0% (by titrimetric analysis)
7-Anilino-4-hydroxy-2-naphthalenesulphonic acid
Formula: C₁₆H₁₃NO₄S
Peso molecolare: 315,34 g/mol
MDL Number: MFCD00035714
CAS Number: 119-40-4
EINECS: 204-320-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR