Stanolone ≥99.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
5alpha-Dihydrotestosterone, DHT, 5alpha-Androstan-17beta-ol-3-one
Formula:
C₁₉H₃₀O₂ Peso molecolare: 290.45 g/mol |
MDL Number:
MFCD00003667 CAS Number: 521-18-6 EINECS: 208-307-3 Merck Index: 14,08793 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 179.0 to 184.0 deg-C |
Specific rotation [a]20/D | +25.0 to +28.0 deg (C=1, Dioxane) |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...