Stampa…

Sodium-7-amino-2-naphthalenesulphonate ≥70% (by total nitrogen basis)

Fornitore: TCI
Attenzione

A0355-25G A0355-500G
TCIAA0355-25GEA 25 EUR
TCIAA0355-25G TCIAA0355-500G
Sodium-7-amino-2-naphthalenesulphonate ≥70% (by total nitrogen basis)
Sodium-7-amino-2-naphthalenesulphonate
Formula: C₁₀H₉NO₃S
MDL Number: MFCD00053436
CAS Number: 5412-82-8
EINECS: 226-498-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4fc009b186acc515ec19e50b454fd334cf8ab0f&vid=b4fc009b186acc515ec19e50b454fd334cf8ab0f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (with Total Nitrogen) min. 70.0 % (calcd.on dried substance)
Drying loss max. 10.0 %
Sodium Chloride max. 25.0 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR