DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
3-Amino-2-methylpropanoic acid, BAIBA
A0324-1G
A0324-5G
TCIAA0324-1GEA
48
EUR
TCIAA0324-1G
TCIAA0324-5G
DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)
DL-3-Aminoisobutyric acid hydrate
Formula:
C₄H₉NO₂·H₂O Peso molecolare: 103,12 g/mol |
MDL Number:
MFCD00008145 CAS Number: 144-90-1 EINECS: 205-644-8 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0% (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | max. 15.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...