Stampa…

Allyl ether ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Diallyl Ether

A0229-25ML A0229-500ML A0229-100ML
TCIAA0229-25MLEA 46 EUR
TCIAA0229-25ML TCIAA0229-500ML TCIAA0229-100ML
Allyl ether ≥98.0% (by GC)
Allyl ether
Formula: C₆H₁₀O
Peso molecolare: 98,15 g/mol
Punto di ebollizione: 94°C
Flash Pt: –7°C
MDL Number: MFCD00008650
CAS Number: 557-40-4
EINECS: 209-174-4
UN: 2360
ADR: 3,II
Merck Index: 14,00292

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR