DL-Adrenaline ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
DL-Epinephrine
A0172-1G
A0172-25G
TCIAA0172-1GEA
24
EUR
TCIAA0172-1G
TCIAA0172-25G
DL-Adrenaline ≥98.0% (by titrimetric analysis)
DL-Adrenaline
Formula:
C₉H₁₃NO₃ Peso molecolare: 183,21 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00063027 CAS Number: 329-65-7 EINECS: 206-347-6 UN: 2811 ADR: 6.1,III Merck Index: 14,03619 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...