Stampa…

Adenine hydrochloride hemihydrate, hemihydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Purin-6-amine hydrochloride hemihydrate, 1H-Purin-6-amine hydrochloride, 7H-Purin-6-amine hydrochloride hemihydrate, 6-Aminopurine Hydrochloride, Adenine Hydrochloride Hemihydrate, 6-Aminopurine hydrochloride hemihydrate, 7H-Purin-6-amine hydrochloride hydrate (2:2:1)

A0150-25G A0150-5G
TCIAA0150-25GEA 97 EUR
TCIAA0150-25G TCIAA0150-5G
Adenine hydrochloride hemihydrate, hemihydrate ≥98.0% (by titrimetric analysis)
Adenine hydrochloride hemihydrate
Formula: C₅H₅N₅·HCl·½H₂O
Peso molecolare: 180.60 g/mol
MDL Number: MFCD00075782
CAS Number: 6055-72-7
EINECS: 220-867-0
Merck Index: 14,00152

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 % (calcd.on dried substance)
Solubility in hot Water almost transparency
Drying loss 4.0 to 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR