Methyl acetylsalicylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl 2-acetoxybenzoate, Methyl 2-Acetoxybenzoate, Aspirin methyl ester, Acetylsalicylic Acid Methyl Ester, 2-Acetoxybenzoic Acid Methyl Ester
A0114-500G
A0114-25G
TCIAA0114-500GEA
134
EUR
TCIAA0114-500G
TCIAA0114-25G
Methyl acetylsalicylate ≥98.0% (by GC)
Methyl acetylsalicylate
Formula:
C₁₀H₁₀O₄ Peso molecolare: 194,19 g/mol |
MDL Number:
MFCD00014978 CAS Number: 580-02-9 EINECS: 209-450-4 Merck Index: 14,06010 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...