Stampa…

Acetone semicarbazone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(Propan-2-ylidene)hydrazinecarboxamide

A0058-25G A0058-500G
TCIAA0058-25GEA 17 EUR
TCIAA0058-25G TCIAA0058-500G
Acetone semicarbazone ≥98.0% (by titrimetric analysis)
Acetone semicarbazone
Formula: C₄H₉N₃O
Peso molecolare: 115,14 g/mol
MDL Number: MFCD00014785
CAS Number: 110-20-3
EINECS: 203-746-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR