Sodium 4-acetamidobenzenesulphinate dihydrate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₈H₉NO₃S |
MDL Number:
MFCD04113633 CAS Number: 15898-43-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Sodium hypochlorite Method) | min. 98.0% (after drying) |
Solubility in Water | almost transparency |
Drying loss | 14.0 to 16.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...