Stampa…

(1S)-(-)-Borneol, white crystalline powder

Fornitore: MP Biomedicals
Pericolo

Synonyms: (-)-Borneol, L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

0215051325 0215051310 0215051380
ICNA0215051325EA 42.3 EUR
ICNA0215051325 ICNA0215051310 ICNA0215051380
(1S)-(-)-Borneol, white crystalline powder
(1S)-(-)-Borneol
Formula: C₁₀H₁₈O
Peso molecolare: 154,25 g/mol
Punto di ebollizione: 203 °C (1013 hPa)
Densità: 1,011 g/cm³ (20 °C)
MDL Number: MFCD00003759
CAS Number: 464-45-9
EINECS: 207-353-1
UN: 1312
ADR: 4.1,III
Merck Index: 13,01328

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR