Calcium iodide hydrate
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
CAS No.: 71626-98-7
Molecular Formula: CaI2.xH2O
Formula Weight: 293.89(anhy)
Physical Form: Lump
Density: 2.55
Sensitivity: Hygroscopic
Appearance: White to grey
Melting Point: 42° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,1677
MDL No.: MFCD00149622
Formula:
CaI₂·H₂O Peso molecolare: 311.9 g/mol Punto di fusione: 42 °C (dec.) Densità: 2.55 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00149622 CAS Number: 71626-98-7 EINECS: 233-276-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...