Stampa…

Etere di petrolio, 40 °C ~, EMPLURA®, Supelco®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ200

1.00915.1000 1.00915.6025
1.00915.1000EA 95 EUR
1.00915.1000 1.00915.6025
Etere di petrolio, 40 °C ~, EMPLURA®, Supelco®
Etere di petrolio
Punto di ebollizione: ∼40 °C (1013 hPa)
Densità: 0,62 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,I

Visualizza i codici

Risultati del test di specificazione

Boiling range (30-40°C) ≥ 90 % (v/v)
Free acid (as CH₃COOH) ≤ 0.0005 %
Density (d 20 °C/ 4 °C) 0.623 - 0.627
Readily carbonizable substances conforms
Evaporation residue ≤ 0.0020 %
Water ≤ 0.010 %
Aromatics (as benzene) ≤ 0.01 %
Sulfur compounds (as S) ≤ 0.01 %
Al (Aluminium) ≤ 0.00002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.0001 %
Cd (Cadmium) ≤ 0.00001 %
Co (Cobalt) ≤ 0.00001 %
Cr (Chromium) ≤ 0.000003 %
Cu (Copper) ≤ 0.000003 %
Fe (Iron) ≤ 0.000003 %
K (Potassium) ≤ 0.00003 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000003 %
Na (Sodium) ≤ 0.0001 %
Ni (Nickel) ≤ 0.000003 %
Pb (Lead) ≤ 0.00001 %
Sr (Strontium) ≤ 0.000003 %
Zn (Zinc) ≤ 0.00003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR